This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407912_m1
See other LINC00601 GE Assays ›
Gene Symbol:
LINC00601
Gene Name:
long intergenic non-protein coding RNA 601
Gene Aliases:
-
Chromosome Location:
Chr.10: 126413869 - 126421879 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407912_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00601
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073453.1 3 - 4 292 98
GenBank mRNA BF215388.1 - 3 - 4 291 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ADAM metallopeptidase domain 12 ADAM12 ADAM12-OT1,
CAR10,
MCMP,
MCMPMltna,
MLTN,
MLTNA
-25.3kb Sense Chr. 10 - 126012385 - 126388558 on NCBI Build GRCh38
Downstream hCG2044975 LOC728158 6.2kb Anti-sense Chr. 10 - 126423107 - 126440535 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012197.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Clariom D Assay PSR1000183375.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000183375.hg.1 NR_073453.1 Refseq
Affymetrix Human Clariom D Assay PSR1000183375.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000183375.hg.1 NR_073453.1 Refseq
Affymetrix Human Clariom D Assay PSR1000183375.hg.1 TC1000012197.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012197.hg.1 ENST00000456514.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Clariom D Assay TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Clariom D Assay TC1000012197.hg.1 NR_073453.1 Refseq
Affymetrix Human Clariom D Assay TC1000012197.hg.1 NR_073453.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline