This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408019_s1
See other YTHDF3-AS1 GE Assays ›
Gene Symbol:
YTHDF3-AS1
Gene Name:
YTHDF3 antisense RNA 1 (head to head)
Gene Aliases:
-
Chromosome Location:
Chr.8: 63167725 - 63168442 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408019_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YTHDF3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102684.1 1 621 97
GenBank mRNA BM967225.1 - 1 76 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW305188.1 - 1 Chr. 8 - 63167723 - 63168108 [+] on Build GRCh38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream alpha tocopherol transfer protein TTPA ATTP,
AVED,
TTP1,
alphaTTP
-81.6kb Sense Chr. 8 - 63058409 - 63086523 on NCBI Build GRCh38
Downstream YTH N6-methyladenosine RNA binding protein 3 YTHDF3 111.0 Anti-sense Chr. 8 - 63168553 - 63212788 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010634.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800010634.hg.1 ENST00000603538.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010634.hg.1 NR_102684.1 Refseq
Affymetrix Human Clariom D Assay TC0800010634.hg.1 ENST00000603538.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010634.hg.1 NR_102684.1 Refseq
Affymetrix Human Clariom D Assay PSR0800172212.hg.1 ENST00000603538.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800172212.hg.1 NR_102684.1 Refseq
Affymetrix Human Clariom D Assay PSR0800172212.hg.1 ENST00000603538.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800172212.hg.1 NR_102684.1 Refseq
Affymetrix Human Clariom D Assay PSR0800172212.hg.1 TC0800010634.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline