This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408721_s1
See other ZBTB46-AS1 GE Assays ›
Gene Symbol:
ZBTB46-AS1
Gene Name:
ZBTB46 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 63808076 - 63823015 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408721_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZBTB46-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110081.1 3 720 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger and BTB domain containing 46 ZBTB46 BTBD4,
BZEL,
RINZF,
ZNF340,
dJ583P15.7,
dJ583P15.8
Anti-sense Chr. 20 - 63743668 - 63832332 on NCBI Build GRCh38
Downstream chromosome 20 open reading frame 181 C20orf181 dJ824A14.1 9.6kb Anti-sense Chr. 20 - 63843262 - 63848312 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-555ccb4fbd-sfw2w:80/100.66.79.196:80.
git-commit: 75f0c7d4cb44686da2af39b21f2e6446a20282e2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-2025.12.99-1.0