This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04702294_m1
See other EIF3M GE Assays ›
Gene Symbol:
EIF3M
Gene Name:
eukaryotic translation initiation factor 3 subunit M
Gene Aliases:
B5, GA17, PCID1, TANGO7, hfl-B5
Chromosome Location:
Chr.11: 32583767 - 32602873 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04702294_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3M
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307929.1 NP_001294858.1 4 - 5 448 74
NM_006360.5 NP_006351.2 7 - 8 844 74
XM_011519840.2 XP_011518142.1 7 - 8 868 74
GenBank mRNA AF064603.1 - 7 - 8 774 74
AF277183.1 - 3 - 4 213 74
AK001768.1 - 3 - 4 1349 74
AK131064.1 - 7 - 8 798 74
AK222564.1 - 7 - 8 763 74
AK292139.1 - 7 - 8 786 74
AK300002.1 - 5 - 6 1628 74
AK304378.1 - 4 - 5 384 74
AK307121.1 - 4 - 5 1464 74
AK312512.1 - 7 - 8 813 74
AV713889.1 - 4 - 5 365 74
AY769947.1 - 7 - 8 723 74
BC019103.2 - 7 - 8 766 5021401 74
BC020458.1 - 3 - 4 235 74
BC051292.1 - 7 - 8 768 3607828 74
CR450300.1 - 7 - 8 723 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3M

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translational initiation
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0