This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04750126_m1
See other SLC35A2 GE Assays ›
Gene Symbol:
SLC35A2
Gene Name:
solute carrier family 35 member A2
Gene Aliases:
CDG2M, CDGX, UDP-Gal-Tr, UGALT, UGAT, UGT, UGT1, UGT2, UGTL
Chromosome Location:
Chr.X: 48903180 - 48911958 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04750126_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC35A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001032289.2 NP_001027460.1 2 - 3 595 77
NM_001042498.2 NP_001035963.1 2 - 3 595 77
NM_001282647.1 NP_001269576.1 2 - 3 595 77
NM_001282648.1 NP_001269577.1 2 - 3 282 77
NM_001282650.1 NP_001269579.1 2 - 3 634 77
NM_001282651.1 NP_001269580.1 3 - 4 679 77
NM_005660.2 NP_005651.1 2 - 3 595 77
GenBank mRNA AK090730.1 - 1 - 2 67 77
AK225508.1 - 2 - 3 283 77
AK292816.1 - 2 - 3 312 77
AK293419.1 - 2 - 3 322 77
AK298484.1 - 3 - 4 367 77
AK299748.1 - 2 - 3 322 77
AL556714.3 - 2 - 3 282 77
BC035747.1 - 2 - 3 293 5749525 77
BI820134.1 - 2 - 3 266 77
CR450287.1 - 2 - 3 273 77
D84454.1 - 2 - 3 596 77
D88146.1 - 2 - 3 277 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC35A2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

galactose metabolic process
UDP-galactose transport
proton transport
UDP-galactose transmembrane transport
sugar:proton symporter activity
UDP-galactose transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0