This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04822952_m1
See other ASIC2 GE Assays ›
Gene Symbol:
ASIC2
Gene Name:
acid sensing ion channel subunit 2
Gene Aliases:
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1
Chromosome Location:
Chr.17: 33013087 - 34156806 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04822952_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASIC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183377.1 NP_899233.1 1 - 2 1580 105
GenBank mRNA AL834182.1 - 1 - 2 1580 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ASIC2

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of systemic arterial blood pressure by aortic arch baroreceptor feedback
chemical synaptic transmission
central nervous system development
peripheral nervous system development
phototransduction
sensory perception of sound
response to acidic pH
regulation of gene expression
monovalent inorganic cation transport
regulation of vasoconstriction
regulation of blood coagulation
ion transmembrane transport
regulation of ion transmembrane transport
protein localization to synapse
sodium ion transmembrane transport
regulation of membrane potential
negative regulation of apoptotic process
sensory perception of sour taste
detection of mechanical stimulus involved in sensory perception
positive regulation of synapse assembly
protein binding
ligand-gated sodium channel activity
ion gated channel activity
acid-sensing ion channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0