This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04935419_m1
See other NEIL2 GE Assays ›
Gene Symbol:
NEIL2
Gene Name:
nei like DNA glycosylase 2
Gene Aliases:
NEH2, NEI2
Chromosome Location:
Chr.8: 11769663 - 11787346 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04935419_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEIL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135748.1 NP_001129220.1 3 - 4 434 68
GenBank mRNA AK294224.1 - 3 - 4 387 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair
depyrimidination
damaged DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
protein binding
microtubule binding
zinc ion binding
DNA N-glycosylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000284503.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000403422.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000436750.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000455213.6 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000524741.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528113.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 ENST00000528323.5 Ensembl
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135746.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135747.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_001135748.2 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq
Affymetrix Human Clariom D Assay TC0800006758.hg.1 NM_145043.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0