This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04935813_g1
See other MSANTD2 GE Assays ›
Gene Symbol:
MSANTD2
Gene Name:
Myb/SANT DNA binding domain containing 2
Gene Aliases:
C11orf61
Chromosome Location:
Chr.11: 124766492 - 124800879 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04935813_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001312919.1 NP_001299848.1 2 - 3 591 129
GenBank mRNA AK303902.1 - 2 - 3 570 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000239614.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000374979.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000524950.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000526629.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000527140.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 ENST00000533514.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_001308027.1 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq
Affymetrix Human Clariom D Assay TC1100012687.hg.1 NM_024631.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0