This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04935835_m1
See other RIPPLY3 GE Assays ›
Gene Symbol:
RIPPLY3
Gene Name:
ripply transcriptional repressor 3
Gene Aliases:
DSCR6
Chromosome Location:
Chr.21: 37006150 - 37019659 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04935835_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIPPLY3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317777.1 NP_001304706.1 2 - 3 103 65
GenBank mRNA AB037160.1 - 2 - 3 103 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB037160.1, NM_001317777.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
heart development
biological_process
negative regulation of cell proliferation
embryonic pattern specification
pharyngeal system development

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000329553.2 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000485272.5 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 ENST00000490393.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Clariom D Assay TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Clariom D Assay TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Clariom D Assay TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Clariom D Assay TC2100007066.hg.1 NM_018962.2 Refseq
Affymetrix Human Clariom D Assay TC2100007066.hg.1 NM_018962.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0