This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04935999_m1
See other PMP2 GE Assays ›
Gene Symbol:
PMP2
Gene Name:
peripheral myelin protein 2
Gene Aliases:
FABP8, M-FABP, MP2, P2
Chromosome Location:
Chr.8: 81440326 - 81447484 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04935999_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PMP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002677.3 NP_002668.1 3 - 4 439 102
GenBank mRNA AK307118.1 - 2 - 3 221 102
AK311758.1 - 3 - 4 394 102
BC034997.1 - 3 - 4 444 4824065 102
CR541649.1 - 3 - 4 341 102
CR541738.1 - 3 - 4 341 102
X62167.1 - 3 - 4 375 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PMP2

Gene Ontology Categories:

Function(s) Process(es)

transport
membrane organization
transporter activity
fatty acid binding
protein binding
cholesterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000256103.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 ENST00000519260.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq
Affymetrix Human Clariom D Assay TC0800010936.hg.1 NM_002677.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline