This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04936104_m1
See other RIMS2 GE Assays ›
Gene Symbol:
RIMS2
Gene Name:
regulating synaptic membrane exocytosis 2
Gene Aliases:
OBOE, RAB3IP3, RIM2
Chromosome Location:
Chr.8: 103500026 - 104254430 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04936104_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011517395.2 XP_011515697.1 2 - 3 229 91
XM_011517398.2 XP_011515700.1 2 - 3 229 91
XM_017014006.1 XP_016869495.1 2 - 3 229 91
XM_017014007.1 XP_016869496.1 2 - 3 229 91
XM_017014008.1 XP_016869497.1 2 - 3 229 91
XM_017014009.1 XP_016869498.1 2 - 3 229 91
XM_017014010.1 XP_016869499.1 2 - 3 229 91
XM_017014011.1 XP_016869500.1 2 - 3 229 91
XM_017014012.1 XP_016869501.1 2 - 3 229 91
XM_017014013.1 XP_016869502.1 2 - 3 229 91
XM_017014015.1 XP_016869504.1 2 - 3 229 91
XM_017014016.1 XP_016869505.1 2 - 3 229 91
XM_017014017.1 XP_016869506.1 2 - 3 229 91
XM_017014019.1 XP_016869508.1 2 - 3 362 91
XM_017014021.1 XP_016869510.1 2 - 3 229 91
XM_017014023.1 XP_016869512.1 2 - 3 229 91
XM_017014024.1 XP_016869513.1 2 - 3 229 91
XM_017014025.1 XP_016869514.1 2 - 3 229 91
XM_017014026.1 XP_016869515.1 2 - 3 503 91
XM_017014027.1 XP_016869516.1 2 - 3 229 91
XM_017014028.1 XP_016869517.1 2 - 3 229 91
XM_017014031.1 XP_016869520.1 2 - 3 229 91
XM_017014032.1 XP_016869521.1 2 - 3 229 91
XM_017014033.1 XP_016869522.1 2 - 3 1071 91
XM_017014039.1 XP_016869528.1 2 - 3 1072 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
spontaneous neurotransmitter secretion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
ion channel binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0