This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04936735_m1
See other TRAK1 GE Assays ›
Gene Symbol:
TRAK1
Gene Name:
trafficking kinesin protein 1
Gene Aliases:
MILT1, OIP106
Chromosome Location:
Chr.3: 42013802 - 42225890 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04936735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042646.2 NP_001036111.1 2 - 3 506 71
NM_001265608.1 NP_001252537.1 2 - 3 506 71
XM_005264956.2 XP_005265013.1 2 - 3 314 71
XM_005264960.3 XP_005265017.1 2 - 3 314 71
XM_005264962.3 XP_005265019.1 2 - 3 314 71
XM_005264963.3 XP_005265020.1 2 - 3 314 71
XM_006713029.2 XP_006713092.1 2 - 3 314 71
XM_006713030.2 XP_006713093.1 2 - 3 314 71
XM_006713031.3 XP_006713094.1 2 - 3 314 71
XM_011533489.2 XP_011531791.1 2 - 3 314 71
XM_017005906.1 XP_016861395.1 4 - 5 927 71
XM_017005909.1 XP_016861398.1 3 - 4 812 71
XM_017005911.1 XP_016861400.1 3 - 4 812 71
XM_017005912.1 XP_016861401.1 2 - 3 314 71
XM_017005913.1 XP_016861402.1 2 - 3 314 71
GenBank mRNA AB028965.1 - 2 - 3 506 71
AK295480.1 - 2 - 3 544 71
AK295848.1 - 2 - 3 413 71
AK316260.1 - 2 - 3 403 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017005909.1, XM_017005911.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
protein O-linked glycosylation
protein targeting
endosome to lysosome transport
protein binding
GABA receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0