This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04936737_m1
See other TRAK1 GE Assays ›
Gene Symbol:
TRAK1
Gene Name:
trafficking kinesin protein 1
Gene Aliases:
MILT1, OIP106
Chromosome Location:
Chr.3: 42013802 - 42225890 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04936737_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042646.2 NP_001036111.1 4 - 5 698 96
NM_001265608.1 NP_001252537.1 4 - 5 698 96
NM_001265609.1 NP_001252538.1 3 - 4 561 96
NM_001265610.1 NP_001252539.1 3 - 4 561 96
NM_014965.4 NP_055780.2 3 - 4 501 96
XM_005264956.2 XP_005265013.1 4 - 5 506 96
XM_005264958.2 XP_005265015.1 3 - 4 561 96
XM_005264960.3 XP_005265017.1 4 - 5 506 96
XM_005264962.3 XP_005265019.1 4 - 5 506 96
XM_005264963.3 XP_005265020.1 4 - 5 506 96
XM_006713029.2 XP_006713092.1 4 - 5 506 96
XM_006713030.2 XP_006713093.1 4 - 5 506 96
XM_006713031.3 XP_006713094.1 4 - 5 506 96
XM_011533489.2 XP_011531791.1 4 - 5 506 96
XM_017005906.1 XP_016861395.1 6 - 7 1119 96
XM_017005907.1 XP_016861396.1 3 - 4 483 96
XM_017005908.1 XP_016861397.1 3 - 4 561 96
XM_017005909.1 XP_016861398.1 5 - 6 1004 96
XM_017005910.1 XP_016861399.1 3 - 4 561 96
XM_017005911.1 XP_016861400.1 5 - 6 1004 96
XM_017005912.1 XP_016861401.1 4 - 5 506 96
XM_017005913.1 XP_016861402.1 4 - 5 506 96
GenBank mRNA AB028965.1 - 4 - 5 698 96
AK000754.1 - 3 - 4 477 96
AK294221.1 - 3 - 4 527 96
AK295480.1 - 4 - 5 736 96
AK295848.1 - 4 - 5 605 96
AK316260.1 - 4 - 5 595 96
AL713787.1 - 3 - 4 465 96
BC015922.1 - 3 - 4 488 4562916 96
BX647199.1 - 3 - 4 501 96
DA193280.1 - 3 - 4 562 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAK1

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
protein O-linked glycosylation
protein targeting
endosome to lysosome transport
protein binding
GABA receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0