This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04936738_m1
See other TRAK1 GE Assays ›
Gene Symbol:
TRAK1
Gene Name:
trafficking kinesin protein 1
Gene Aliases:
MILT1, OIP106
Chromosome Location:
Chr.3: 42013802 - 42225890 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04936738_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
4,430.00
Your Price
Online offer:
4430.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TRAK1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042646.2
NP_001036111.1
5 - 6
800
58
NM_001265608.1
NP_001252537.1
5 - 6
800
58
NM_001265609.1
NP_001252538.1
4 - 5
663
58
NM_001265610.1
NP_001252539.1
4 - 5
663
58
NM_014965.4
NP_055780.2
4 - 5
603
58
XM_005264956.2
XP_005265013.1
5 - 6
608
58
XM_005264958.2
XP_005265015.1
4 - 5
663
58
XM_005264960.3
XP_005265017.1
5 - 6
608
58
XM_005264962.3
XP_005265019.1
5 - 6
608
58
XM_005264963.3
XP_005265020.1
5 - 6
608
58
XM_006713029.2
XP_006713092.1
5 - 6
608
58
XM_006713030.2
XP_006713093.1
5 - 6
608
58
XM_006713031.3
XP_006713094.1
5 - 6
608
58
XM_011533489.2
XP_011531791.1
5 - 6
608
58
XM_017005906.1
XP_016861395.1
7 - 8
1221
58
XM_017005907.1
XP_016861396.1
4 - 5
585
58
XM_017005908.1
XP_016861397.1
4 - 5
663
58
XM_017005909.1
XP_016861398.1
6 - 7
1106
58
XM_017005910.1
XP_016861399.1
4 - 5
663
58
XM_017005911.1
XP_016861400.1
6 - 7
1106
58
XM_017005912.1
XP_016861401.1
5 - 6
608
58
XM_017005913.1
XP_016861402.1
5 - 6
608
58
GenBank mRNA
AB028965.1
-
5 - 6
800
58
AK000754.1
-
4 - 5
579
58
AK294221.1
-
4 - 5
629
58
AK295480.1
-
5 - 6
838
58
AK295848.1
-
5 - 6
707
58
AK316260.1
-
5 - 6
697
58
AL713787.1
-
4 - 5
567
58
BC015922.1
-
4 - 5
590
4562916
58
BX647199.1
-
4 - 5
603
58
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene TRAK1
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
protein O-linked glycosylation
protein targeting
endosome to lysosome transport
protein binding
GABA receptor binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0