Assay Details
Gene Symbol: HSD17B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304512.1 | NP_001291441.1 | 4 | 625 | 92 | |
NM_001304513.1 | NP_001291442.1 | 4 | 599 | 92 | ||
GenBank mRNA | AK022929.1 | - | 4 | 625 | 92 | |
AW136392.1 | - | 1 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000488656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000488656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000488656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
