This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937837_m1
See other LOC105378732 GE Assays ›
Gene Symbol:
LOC105378732
Gene Name:
uncharacterized LOC105378732
Gene Aliases:
-
Chromosome Location:
Chr.1: 53328233 - 53336509 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937837_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105378732
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_131923.1 1 - 2 140 153
GenBank mRNA AK097571.1 - 1 - 2 141 153
DB038107.1 - 1 - 2 141 153

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream LDL receptor related protein 8 LRP8 APOER2,
HSZ75190,
LRP-8,
MCI1
-84.0 Anti-sense Chr. 1 - 53242364 - 53328149 on NCBI Build GRCh38
Downstream solute carrier family 25 member 3 pseudogene 1 SLC25A3P1 107.1kb Anti-sense Chr. 1 - 53438370 - 53440020 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008356.hg.1 ENST00000445039.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008356.hg.1 ENST00000445039.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008356.hg.1 ENST00000445039.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008356.hg.1 NR_131923.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008356.hg.1 NR_131923.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008356.hg.1 NR_131923.1 Refseq
Affymetrix Human Clariom D Assay TC0100008356.hg.1 ENST00000445039.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008356.hg.1 ENST00000445039.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008356.hg.1 ENST00000445039.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008356.hg.1 NR_131923.1 Refseq
Affymetrix Human Clariom D Assay TC0100008356.hg.1 NR_131923.1 Refseq
Affymetrix Human Clariom D Assay TC0100008356.hg.1 NR_131923.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline