This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938162_m1
See other LOC105378349 GE Assays ›
Gene Symbol:
LOC105378349
Gene Name:
uncharacterized LOC105378349
Gene Aliases:
-
Chromosome Location:
Chr.10: 70939053 - 70955641 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938162_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105378349
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134313.1 2 - 3 462 79
GenBank mRNA DB069022.1 - 2 - 3 463 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream pterin-4 alpha-carbinolamine dehydratase 1 PCBD1 DCOH,
PCBD,
PCD,
PHS
-65.4kb Anti-sense Chr. 10 - 70882280 - 70888786 on NCBI Build GRCh38
Downstream unc-5 netrin receptor B UNC5B UNC5H2,
p53RDL1
256.9kb Sense Chr. 10 - 71212535 - 71302879 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000558852.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000560700.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007942.hg.1 ENST00000560700.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline