This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938253_m1
See other LOC105378098 GE Assays ›
Gene Symbol:
LOC105378098
Gene Name:
uncharacterized LOC105378098
Gene Aliases:
-
Chromosome Location:
Chr.6: 161470977 - 161484045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938253_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105378098
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134593.1 1 - 2 44 98
NR_134594.1 1 - 2 44 98
NR_134595.1 1 - 2 44 98
GenBank mRNA DB041402.1 - 1 - 2 44 98
DB083190.1 - 1 - 2 44 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream 1-acylglycerol-3-phosphate O-acyltransferase 4 AGPAT4 1-AGPAT4,
LPAAT-delta,
dJ473J16.2
-196.9kb Anti-sense Chr. 6 - 161129980 - 161274108 on NCBI Build GRCh38
Downstream PARK2 coregulated PACRG GLUP,
HAK005771,
PARK2CRG
1.2mb Sense Chr. 6 - 162726870 - 163315492 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0