This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938458_m1
See other LOC105369632 GE Assays ›
Gene Symbol:
LOC105369632
Gene Name:
uncharacterized LOC105369632
Gene Aliases:
-
Chromosome Location:
Chr.12: 6875764 - 6889358 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938458_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105369632
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135083.1 2 - 3 155 70
GenBank mRNA AI674052.1 - 2 - 3 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX331561.2 - 1 - 2 104 Chr. 12 - 6872655 - 6882289 [-] on Build GRCh38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream splA/ryanodine receptor domain and SOCS box containing 2 SPSB2 GRCC9,
SSB2
-4.1kb Sense Chr. 12 - 6870935 - 6873357 on NCBI Build GRCh38
Downstream ribosomal protein L13 pseudogene 5 RPL13P5 RPL13-2,
RPL13L,
RPL13_4_1199,
RRPL13L
1.7kb Anti-sense Chr. 12 - 6883981 - 6884614 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012743.hg.1 ENST00000437851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012743.hg.1 ENST00000437851.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline