This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938475_m1
See other LOC105369509 GE Assays ›
Gene Symbol:
LOC105369509
Gene Name:
uncharacterized LOC105369509
Gene Aliases:
-
Chromosome Location:
Chr.11: 115333577 - 115340439 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938475_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105369509
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135108.1 1 - 2 33 92
GenBank mRNA BX119732.1 - 1 - 2 33 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neurexophilin and PC-esterase domain family member 2 NXPE2 FAM55B -519.9kb Sense Chr. 11 - 114464276 - 114813702 on NCBI Build GRCh38
Downstream uncharacterized LOC105369507 LOC105369507 56.3kb Sense Chr. 11 - 115396756 - 115397658 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009127.hg.1 ENST00000545593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009127.hg.1 ENST00000545593.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009127.hg.1 ENST00000545593.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009127.hg.1 ENST00000545593.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline