This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938598_m1
See other LOC101929054 GE Assays ›
Gene Symbol:
LOC101929054
Gene Name:
uncharacterized LOC101929054
Gene Aliases:
-
Chromosome Location:
Chr.3: 52239238 - 52241097 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938598_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101929054
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135307.1 1 - 2 198 69
GenBank mRNA BC039681.1 - 1 - 2 172 69
HY101534.1 - 1 - 2 198 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA934269.1 - 1 - 2 180 Chr. 3 - 52239258 - 52240479 [+] on Build GRCh38 69
GenBankmRNA BI911063.1 - 1 - 2 172 Chr. 3 - 52239264 - 52240912 [+] on Build GRCh38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream twinfilin actin binding protein 2 TWF2 A6RP,
A6r,
MSTP011,
PTK9L
-71.0 Anti-sense Chr. 3 - 52228610 - 52239167 on NCBI Build GRCh38
Downstream protein phosphatase,
Mg2+/Mn2+ dependent 1M
PPM1M PP2C-eta,
PP2CE,
PP2Ceta
4.6kb Sense Chr. 3 - 52245725 - 52250599 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300007468.hg.1 ENST00000464958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007468.hg.1 ENST00000464958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007468.hg.1 ENST00000483834.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007468.hg.1 ENST00000483834.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007468.hg.1 ENST00000483834.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007468.hg.1 ENST00000464958.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007468.hg.1 ENST00000464958.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007468.hg.1 ENST00000483834.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007468.hg.1 ENST00000483834.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007468.hg.1 ENST00000483834.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0