This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938664_m1
See other LOC101926983 GE Assays ›
Gene Symbol:
LOC101926983
Gene Name:
uncharacterized LOC101926983
Gene Aliases:
-
Chromosome Location:
Chr.3: 120812181 - 120836357 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938664_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101926983
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135570.1 1 - 2 79 93
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI829396.1 - 1 - 2 67 Chr. 3 - 120811835 - 120836345 [-] on Build GRCh38 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream general transcription factor IIE subunit 1 GTF2E1 FE,
TF2E1,
TFIIE-A
-29.0kb Anti-sense Chr. 3 - 120742711 - 120783134 on NCBI Build GRCh38
Downstream syntaxin binding protein 5 like STXBP5L LLGL4 71.8kb Anti-sense Chr. 3 - 120908203 - 121424761 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012158.hg.1 ENST00000490647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012158.hg.1 ENST00000490647.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012158.hg.1 ENST00000490647.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012158.hg.1 ENST00000490647.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0