This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938670_g1
See other DPY30 GE Assays ›
Gene Symbol:
DPY30
Gene Name:
dpy-30, histone methyltransferase complex regulatory subunit
Gene Aliases:
Cps25, HDPY-30, Saf19
Chromosome Location:
Chr.2: 32011649 - 32039840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938670_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DPY30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135587.1 2 - 3 178 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mediator of cell motility 1 MEMO1 C2orf4,
CGI-27,
MEMO,
NS5ATP7
-28.2kb Sense Chr. 2 - 31867188 - 32011052 on NCBI Build GRCh38
Downstream spastin SPAST ADPSP,
FSP2,
SPG4
24.1kb Anti-sense Chr. 2 - 32063551 - 32157637 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at telomere
transcription, DNA-templated
endosomal transport
histone H3-K4 methylation
protein binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline