This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938810_m1
See other HDAC9 GE Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938810_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135835.1 2 - 3 379 64
GenBank mRNA BC092441.1 - 2 - 3 350 64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD522808.1 - 2 - 3 353 Chr. 7 - 18495739 - 18548404 [+] on Build GRCh38 64
GenBankmRNA DA783769.1 - 2 - 3 550 Chr. 7 - 18495540 - 18543204 [+] on Build GRCh38 64
GenBankmRNA CD684529.1 - 2 - 3 397 Chr. 7 - 18495739 - 18543404 [+] on Build GRCh38 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1302-6 MIR1302-6 MIRN1302-6,
hsa-mir-1302-6
-369.0kb Anti-sense Chr. 7 - 18127220 - 18127309 on NCBI Build GRCh38
Downstream twist family bHLH transcription factor 1 TWIST1 ACS3,
BPES2,
BPES3,
CRS,
CRS1,
CSO,
SCS,
TWIST,
bHLHa38
569.8kb Anti-sense Chr. 7 - 19113047 - 19117672 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline