Assay Details
| Gene Symbol: HDAC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135835.1 | 3 - 4 | 497 | 96 | ||
| GenBank mRNA | BC092441.1 | - | 3 - 4 | 468 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1302-6 | MIR1302-6 |
MIRN1302-6, hsa-mir-1302-6 |
-415.9kb | Anti-sense | Chr. 7 - 18127220 - 18127309 on NCBI Build GRCh38 |
| Downstream | twist family bHLH transcription factor 1 | TWIST1 |
ACS3, BPES2, BPES3, CRS, CRS1, CSO, SCS, TWIST, bHLHa38 |
564.5kb | Anti-sense | Chr. 7 - 19113047 - 19117672 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

HDAC9
Genomic Map