This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938870_m1
See other LOC101928166 GE Assays ›
Gene Symbol:
LOC101928166
Gene Name:
uncharacterized LOC101928166
Gene Aliases:
-
Chromosome Location:
Chr.3: 152159464 - 152205429 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938870_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101928166
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136178.1 1 - 2 56 85
GenBank mRNA KR024014.1 - 2 - 3 85
KR024015.1 - 1 - 2 56 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream AADACL2 antisense RNA 1 AADACL2-AS1 -246.6kb Sense Chr. 3 - 151751179 - 151928175 on NCBI Build GRCh38
Downstream muscleblind like splicing regulator 1 MBNL1 EXP,
MBNL
38.2kb Anti-sense Chr. 3 - 152243656 - 152465780 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline