This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04939065_m1
See other DSE GE Assays ›
Gene Symbol:
DSE
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
Chromosome Location:
Chr.6: 116254152 - 116441261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04939065_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136522.1 1 - 2 194 95
XM_017010795.1 XP_016866284.1 1 - 2 71 95
GenBank mRNA BX110619.1 - 1 - 2 102 95
DC333659.1 - 1 - 2 194 95
HY012067.1 - 1 - 2 159 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DC416992.1 - 1 - 2 171 Chr. 6 - 116280086 - 116399276 [+] on Build GRCh38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TSPY like 4 TSPYL4 dJ486I3.2 -26.0kb Anti-sense Chr. 6 - 116249964 - 116254098 on NCBI Build GRCh38
Downstream uncharacterized LOC100287467 LOC100287467 1.3kb Anti-sense Chr. 6 - 116368563 - 116372237 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017010795.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
chondroitin sulfate biosynthetic process
dermatan sulfate biosynthetic process
chondroitin-glucuronate 5-epimerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000331677.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000359564.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000449314.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000452085.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000453463.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606179.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606265.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606265.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606265.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000606712.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 ENST00000607094.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_001080976.2 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq
Affymetrix Human Clariom D Assay TC0600014177.hg.1 NM_013352.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-llv4q:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0