This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04940077_g1
See other NRM GE Assays ›
Gene Symbol:
NRM
Gene Name:
nurim (nuclear envelope membrane protein)
Gene Aliases:
NRM29
Chromosome Location:
Chr.6: 30688047 - 30691420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04940077_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NRM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073072.1 1 - 2 132 83
GenBank mRNA BE548635.1 - 1 - 2 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein phosphatase 1 regulatory subunit 18 PPP1R18 HKMT1098,
KIAA1949
-2.2kb Sense Chr. 6 - 30676389 - 30687895 on NCBI Build GRCh38
Downstream mediator of DNA damage checkpoint 1 MDC1 NFBD1 9.1kb Sense Chr. 6 - 30699807 - 30717988 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000259953.8 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376420.9 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000376421.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000444096.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000462857.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000470733.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000474864.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000482141.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 ENST00000495946.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_001270707.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NM_007243.2 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073065.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073072.1 Refseq
Affymetrix Human Clariom D Assay TC0600011377.hg.1 NR_073072.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0