This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04940459_gH
See other RNVU1-18 GE Assays ›
Gene Symbol:
RNVU1-18 GSTM2 RNU1-1 RNU1-2 RNU1-3 RNU1-4
Gene Name:
RNA, variant U1 small nuclear 18
glutathione S-transferase mu 2 (muscle)
RNA, U1 small nuclear 1
RNA, U1 small nuclear 2
RNA, U1 small nuclear 3
RNA, U1 small nuclear 4
Gene Aliases:
RNU1-25, RNU1-25P, RNU1-5, RNU1P1, RNU1P9, U1.15, U1P101, U1P15, vU1.18
GST4, GSTM, GSTM2-2, GTHMUS
HSD1, HU1-1, RNU1, RNU1A, RNU1A3, RNU1G4, Rnu1a1, U1, U1A1
RNU1C1, RNU1C2, U1C1, U1C21
HSD4, RNU1G3
HSD2, HSD5, HSD6, HSD7, RNU1, RNU1D2, RNU1E2, RNU1F1, RNU1G1, RNU1G2, U1B2, U1D2, U1E2, U1F
Chromosome Location:
Chr.1: 143729407 - 143729570 on Build GRCh38
Chromosome Location:
Chr.1: 109668022 - 109683997 on Build GRCh38
Chromosome Location:
Chr.1: 16514122 - 16514285 on Build GRCh38
Chromosome Location:
Chr.1: 16895980 - 16896143 on Build GRCh38
Chromosome Location:
Chr.1: 16666785 - 16666948 on Build GRCh38
Chromosome Location:
Chr.1: 16740516 - 16740679 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
-
-
Assay ID Hs04940459_gH
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNVU1-18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004400.1 1 122 74
GenBank mRNA JQ988812.1 - 1 122 74
Gene Symbol: GSTM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK299482.1 - 122 74
Gene Symbol: RNU1-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004430.2 1 122 74
Gene Symbol: RNU1-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004427.1 1 122 74
Gene Symbol: RNU1-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004408.1 1 122 74
Gene Symbol: RNU1-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004421.1 1 122 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA312837.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA713933.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB016209.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC393960.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA421431.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA334550.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA701928.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA810198.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA759737.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA AV737386.1 - 1 167 Chr. 1 - 146376798 - 146377226 [+] on Build GRCh38 74
GenBankmRNA DA124025.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA403529.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA672157.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA838076.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA288321.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DB164756.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA155276.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA536983.1 - 121 Chr. - - [-] on Build 74
GenBankmRNA DA502057.1 - 1 122 Chr. 1 - 244694384 - 245134090 [-] on Build GRCh38 74
GenBankmRNA DC428558.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA440966.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA708812.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DW434924.1 - 1 139 Chr. 14 - 34546714 - 34546953 [+] on Build GRCh38 74
GenBankmRNA DA496966.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB018032.1 - 1 123 Chr. 1 - 96225902 - 96447278 [+] on Build GRCh38 74
GenBankmRNA DA704951.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA BG474191.1 - 1 291 Chr. 1|NT_187368.1 - 21722 - 22193 [-] on Build GRCh38 74
GenBankmRNA AU136324.2 - 114 Chr. - - [-] on Build 74
GenBankmRNA DB083818.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA677551.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA704927.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA263428.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA006206.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DW432578.1 - 1 139 Chr. 14 - 34546714 - 34546960 [+] on Build GRCh38 74
GenBankmRNA DA768895.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA264388.1 - 1 123 Chr. 1 - 96225900 - 96447252 [+] on Build GRCh38 74
GenBankmRNA DB155769.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA518770.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC325985.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA AU138351.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA004725.1 - 1 122 Chr. 1 - 96225902 - 96447179 [+] on Build GRCh38 74
GenBankmRNA DB191411.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA537459.1 - 1 122 Chr. 1 - 96225902 - 96447250 [+] on Build GRCh38 74
GenBankmRNA DA943896.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC419275.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA618097.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA336951.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA002557.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA938507.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA CB131782.1 - 118 Chr. - - [-] on Build 74
GenBankmRNA DA579359.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA578078.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB013631.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC330228.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA619701.1 - 118 Chr. - - [-] on Build 74
GenBankmRNA DA183623.1 - 124 Chr. - - [-] on Build 74
GenBankmRNA DA465028.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA BJ997093.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA971361.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DW432579.1 - 1 139 Chr. 14 - 34546714 - 34546961 [+] on Build GRCh38 74
GenBankmRNA DA387365.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DA257000.1 - 1 122 Chr. 1 - 96225902 - 96447252 [+] on Build GRCh38 74
GenBankmRNA DC326915.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA371669.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC339808.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA372317.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA CA455449.1 - 1 107 Chr. 14 - 34546729 - 34547030 [+] on Build GRCh38 74
GenBankmRNA DB175733.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB192650.1 - 1 118 Chr. 1 - 96225905 - 96447275 [+] on Build GRCh38 74
GenBankmRNA DA808663.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB009629.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA761037.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA257383.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA261098.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB188969.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA677674.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA425312.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA674820.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA172999.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA675443.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA280044.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA252095.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA322830.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DA146370.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA423773.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA333204.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA AU122692.2 - 1 122 Chr. 1 - 96225902 - 96447574 [+] on Build GRCh38 74
GenBankmRNA DA768151.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA152706.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA162073.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA088423.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA674514.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA466951.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA697974.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA739496.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA828081.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DA427841.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA925487.1 - 1 122 Chr. 1 - 96225902 - 96447270 [+] on Build GRCh38 74
GenBankmRNA DA277774.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DW415596.1 - 1 139 Chr. 14 - 34546714 - 34547008 [+] on Build GRCh38 74
GenBankmRNA DB018496.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA671158.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA809216.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC371940.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB198989.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA829038.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DA579268.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB169971.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA878149.1 - 118 Chr. - - [-] on Build 74
GenBankmRNA DA829728.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA260774.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB155802.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA BM749651.1 - 133 Chr. - - [-] on Build 74
GenBankmRNA DA426245.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA742486.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC328436.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA494866.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA BM838247.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA706188.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA791264.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA443352.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB072544.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA267808.1 - 1 122 Chr. 1 - 244694379 - 245134090 [-] on Build GRCh38 74
GenBankmRNA DC356301.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA677525.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA717155.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC341115.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA706746.1 - Chr. - - [-] on Build 74
GenBankmRNA DA781958.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB068203.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB187990.1 - 1 122 Chr. 1 - 96225902 - 96447280 [+] on Build GRCh38 74
GenBankmRNA DA453155.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA768339.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA770561.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA474743.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA676309.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA379111.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB193984.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB246189.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA180975.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA671559.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA703257.1 - 1 122 Chr. 1 - 96225902 - 96447208 [+] on Build GRCh38 74
GenBankmRNA DB018678.1 - 1 122 Chr. 1 - 96225902 - 96447244 [+] on Build GRCh38 74
GenBankmRNA DA677548.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA674754.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA275092.1 - 1 122 Chr. 1 - 96225902 - 96447252 [+] on Build GRCh38 74
GenBankmRNA CB123444.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA618580.1 - 1 122 Chr. 1 - 96225902 - 96447283 [+] on Build GRCh38 74
GenBankmRNA DA751841.1 - 119 Chr. - - [-] on Build 74
GenBankmRNA AV760461.1 - 1 171 Chr. 14 - 34546714 - 34547131 [+] on Build GRCh38 74
GenBankmRNA DA152744.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA752254.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA365716.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA153063.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA BP379965.1 - 222 Chr. - - [-] on Build 74
GenBankmRNA DA950877.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA937797.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA363799.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DA748382.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA677605.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA365019.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA AV738075.1 - 1 183 Chr. 14 - 34546714 - 34547268 [+] on Build GRCh38 74
GenBankmRNA DA953771.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB018102.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA503373.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB192452.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA878181.1 - 1 122 Chr. 1 - 16369147 - 16514284 [-] on Build GRCh38 74
GenBankmRNA DA483459.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA502222.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC387010.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA170710.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA J00318.3 - 1 554 Chr. 1 - 16740084 - 16896354 [+] on Build GRCh38 74
GenBankmRNA DC393923.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC360354.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB232154.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA158185.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB259233.1 - 115 Chr. - - [-] on Build 74
GenBankmRNA DA893735.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA903274.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA947953.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA919174.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA CB108834.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA770456.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA439999.1 - 124 Chr. - - [-] on Build 74
GenBankmRNA DA408970.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA702120.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB243986.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA338407.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA448971.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA325308.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC369735.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA445801.1 - 119 Chr. - - [-] on Build 74
GenBankmRNA DA756749.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA832664.1 - 123 Chr. - - [-] on Build 74
GenBankmRNA DA797548.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA452649.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA674353.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB011107.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB226193.1 - 91 Chr. - - [-] on Build 74
GenBankmRNA AI739023.1 - 1 281 Chr. 1 - 143729175 - 143729511 [+] on Build GRCh38 74
GenBankmRNA DA162108.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA716039.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA760159.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA384883.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA HY019711.1 - 1 366 Chr. 1 - 16895737 - 16896222 [+] on Build GRCh38 74
GenBankmRNA DA676440.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC393348.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA325389.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA191008.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC413511.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DA779316.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DC367150.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA DB245851.1 - 122 Chr. - - [-] on Build 74
GenBankmRNA AL705609.1 - 1 344 Chr. 1|NT_187368.1 - 21779 - 22261 [-] on Build GRCh38 74
GenBankmRNA DA682211.1 - 122 Chr. - - [-] on Build 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC107985108 LOC107985108 Anti-sense Chr. 1 - 143729544 - 143737130 on NCBI Build GRCh38
Downstream uncharacterized LOC102724558 CH17-408M7.1 109.1kb Sense Chr. 1 - 146486332 - 146525289 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK299482.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline