This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04941969_g1
See other LINC01480 GE Assays ›
Gene Symbol:
LINC01480
Gene Name:
long intergenic non-protein coding RNA 1480
Gene Aliases:
-
Chromosome Location:
Chr.19: 41535498 - 41536904 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04941969_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01480
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110724.1 1 - 2 103 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream prostate cancer associated transcript 19 (non-protein coding) PCAT19 LINC01190 -34.8kb Anti-sense Chr. 19 - 41454169 - 41500649 on NCBI Build GRCh38
Downstream carcinoembryonic antigen related cell adhesion molecule 21 CEACAM21 CEACAM3,
R29124_1
13.7kb Sense Chr. 19 - 41549518 - 41586844 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000494375.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000494375.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000494375.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000483481.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000487420.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000494375.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000494375.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000494375.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008155.hg.1 ENST00000597199.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-q45k6:80/100.66.78.42:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0