This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942118_g1
See other LOC101927481 GE Assays ›
Gene Symbol:
LOC101927481
Gene Name:
uncharacterized LOC101927481
Gene Aliases:
-
Chromosome Location:
Chr.18: 70335439 - 70352459 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942118_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927481
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126336.1 1 - 2 66 112
GenBank mRNA AV726231.1 - 1 - 2 112

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream suppressor of cytokine signaling 6 SOCS6 CIS-4,
CIS4,
HSPC060,
SOCS-4,
SOCS-6,
SOCS4,
SSI4,
STAI4,
STATI4
-5.2kb Sense Chr. 18 - 70288901 - 70330199 on NCBI Build GRCh38
Downstream uncharacterized LOC101060542 LOC101060542 44.0kb Anti-sense Chr. 18 - 70380140 - 70384591 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000583247.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 ENST00000584919.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Clariom D Assay TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Clariom D Assay TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Clariom D Assay TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Clariom D Assay TC1800007622.hg.1 NR_126336.1 Refseq
Affymetrix Human Clariom D Assay TC1800007622.hg.1 NR_126336.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline