This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942255_m1
See other P3H2-AS1 GE Assays ›
Gene Symbol:
P3H2-AS1
Gene Name:
P3H2 antisense RNA 1
Gene Aliases:
LEPREL1-AS1
Chromosome Location:
Chr.3: 190120964 - 190144846 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942255_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P3H2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126419.1 2 - 3 269 117
GenBank mRNA BM017065.1 - 2 - 3 270 117

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream prolyl 3-hydroxylase 2 P3H2 LEPREL1,
MCVD,
MLAT4
-6.5kb Anti-sense Chr. 3 - 189956728 - 190122437 on NCBI Build GRCh38
Downstream claudin 1 CLDN1 CLD1,
ILVASC,
SEMP1
162.2kb Anti-sense Chr. 3 - 190305701 - 190322446 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009848.hg.1 ENST00000412203.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Clariom D Assay TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Clariom D Assay TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Clariom D Assay TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Clariom D Assay TC0300009848.hg.1 NR_126419.1 Refseq
Affymetrix Human Clariom D Assay TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008995.hg.1 ENST00000411856.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008995.hg.1 ENST00000411856.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline