This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942275_m1
See other PCAT29 GE Assays ›
Gene Symbol:
PCAT29
Gene Name:
prostate cancer associated transcript 29 (non-protein coding)
Gene Aliases:
-
Chromosome Location:
Chr.15: 69592200 - 69695750 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942275_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCAT29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126438.1 3 - 4 379 105
GenBank mRNA AA635443.1 - 3 - 4 374 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream downregulated RNA in cancer,
inhibitor of cell invasion and migration
DRAIC -106.1kb Sense Chr. 15 - 69561720 - 69571440 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 593 LINC00593 C15orf50 139.5kb Sense Chr. 15 - 69835234 - 69842967 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline