This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942353_m1
See other C19ORF84 GE Assays ›
Gene Symbol:
C19orf84
Gene Name:
chromosome 19 open reading frame 84
Gene Aliases:
-
Chromosome Location:
Chr.19: 51388289 - 51390574 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942353_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C19orf84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126530.1 1 - 2 223 89
GenBank mRNA BC143997.1 - 1 - 2 212 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream lens intrinsic membrane protein 2 LIM2 CTRCT19,
MP17,
MP19
-333.0 Sense Chr. 19 - 51379909 - 51387956 on NCBI Build GRCh38
Downstream sialic acid binding Ig like lectin 10 SIGLEC10 PRO940,
SIGLEC-10,
SLG2
19.4kb Sense Chr. 19 - 51410021 - 51417803 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000570516.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 ENST00000574814.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NM_001193623.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NR_126530.1 Refseq
Affymetrix Human Clariom D Assay TC1900011253.hg.1 NR_126530.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline