This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04962570_m1
See other ATP5L GE Assays ›
Gene Symbol:
ATP5L
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit G
Gene Aliases:
ATP5JG
Chromosome Location:
Chr.11: 118401389 - 118409847 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04962570_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033759.1 2 - 3 386 98
GenBank mRNA BU953217.1 - 2 - 3 171 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
mitochondrial ATP synthesis coupled proton transport
hydrogen ion transmembrane transporter activity
ATPase activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000300688.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000527186.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529460.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529770.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000529790.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000533172.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 ENST00000534385.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NM_006476.4 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq
Affymetrix Human Clariom D Assay TC1100013100.hg.1 NR_033759.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0