This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04962805_m1
See other YPEL1 GE Assays ›
Gene Symbol:
YPEL1
Gene Name:
yippee like 1
Gene Aliases:
FKSG3
Chromosome Location:
Chr.22: 21697536 - 21735834 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04962805_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YPEL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013313.4 NP_037445.1 1 - 2 221 114
NR_130910.1 1 - 2 221 114
GenBank mRNA AB098734.1 - 1 - 2 221 114
AB098735.1 - 1 - 2 221 114
BC034486.1 - 1 - 2 79 4826997 114
BC074501.1 - 1 - 2 86 5171826 114
BI461654.1 - 1 - 2 273 114
BI559478.1 - 1 - 2 101 114

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB098734.1, AB098735.1, BC034486.1, BC074501.1, NM_013313.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene YPEL1

Gene Ontology Categories:

Process(es)

metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000339468.7 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000403503.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 ENST00000477675.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NM_013313.4 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq
Affymetrix Human Clariom D Assay TC2200008103.hg.1 NR_130910.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline