This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04963025_m1
See other ASCC2 GE Assays ›
Gene Symbol:
ASCC2
Gene Name:
activating signal cointegrator 1 complex subunit 2
Gene Aliases:
ASC1p100, p100
Chromosome Location:
Chr.22: 29788608 - 29838304 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04963025_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASCC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011530450.2 XP_011528752.1 4 - 5 379 120
XM_011530451.2 XP_011528753.1 5 - 6 453 120
XM_011530452.2 XP_011528754.1 4 - 5 426 120
XM_011530453.2 XP_011528755.1 3 - 4 281 120
XM_017029005.1 XP_016884494.1 4 - 5 379 120
XM_017029006.1 XP_016884495.1 5 - 6 453 120
XM_017029007.1 XP_016884496.1 3 - 4 281 120
GenBank mRNA AK022886.1 - 4 - 5 341 120

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK022886.1, XM_011530450.2, XM_011530451.2, XM_011530452.2, XM_011530453.2, XM_017029005.1, XM_017029006.1, XM_017029007.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s)

DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline