This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04963506_m1
See other SIAH1 GE Assays ›
Gene Symbol:
SIAH1
Gene Name:
siah E3 ubiquitin protein ligase 1
Gene Aliases:
SIAH1A
Chromosome Location:
Chr.16: 48360531 - 48448440 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04963506_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIAH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011523279.1 XP_011521581.1 2 - 3 1199 79
GenBank mRNA BC044920.1 - 2 - 3 1162 5295925 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC044920.1, XM_011523279.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
ubiquitin-dependent protein catabolic process
apoptotic process
cell cycle
spermatogenesis
nervous system development
axon guidance
anatomical structure morphogenesis
protein catabolic process
protein destabilization
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cellular protein metabolic process
neuron apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
protein C-terminus binding
zinc ion binding
ligase activity
identical protein binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0