This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04964022_s1
See other TRAPPC2L GE Assays ›
Gene Symbol:
TRAPPC2L
Gene Name:
trafficking protein particle complex 2-like
Gene Aliases:
HSPC176
Chromosome Location:
Chr.16: 88856220 - 88862686 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04964022_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC2L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318524.1 NP_001305453.1 5 1662 102
NM_001318525.1 NP_001305454.1 5 1641 102
NM_001318526.1 NP_001305455.1 5 1706 102
NM_001318527.1 NP_001305456.1 5 1978 102
NM_001318528.1 NP_001305457.1 5 1621 102
NM_001318529.1 NP_001305458.1 5 1960 102
NM_001318530.1 NP_001305459.1 5 1618 102
NM_001318532.1 NP_001305461.1 5 1957 102
NM_016209.4 NP_057293.1 5 1644 102
NR_134670.1 6 1828 102
NR_134671.1 6 1760 102
XM_017023270.1 XP_016878759.1 5 1688 102
GenBank mRNA AK126779.1 - 4 2611 102
BC011369.1 - 5 1588 3868798 102
BC017440.1 - 1 919 102
BM974507.1 - 1 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAPPC2L

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0