This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04964695_m1
See other CDH3 GE Assays ›
Gene Symbol:
CDH3
Gene Name:
cadherin 3
Gene Aliases:
CDHP, HJMD, PCAD
Chromosome Location:
Chr.16: 68644820 - 68727573 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04964695_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011522800.2 XP_011521102.1 16 - 17 3033 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
visual perception
positive regulation of gene expression
positive regulation of keratinocyte proliferation
single organismal cell-cell adhesion
hair cycle process
keratinization
positive regulation of monophenol monooxygenase activity
negative regulation of transforming growth factor beta2 production
adherens junction organization
wound healing
response to drug
positive regulation of insulin-like growth factor receptor signaling pathway
positive regulation of melanin biosynthetic process
negative regulation of catagen
canonical Wnt signaling pathway
regulation of hair cycle by canonical Wnt signaling pathway
positive regulation of melanosome transport
molecular_function
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline