This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04965355_g1
See other ATP5G1 GE Assays ›
Gene Symbol:
ATP5G1
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9)
Gene Aliases:
ATP5A, ATP5G
Chromosome Location:
Chr.17: 48892786 - 48895871 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04965355_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5G1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002027.1 NP_001002027.1 2 - 3 102 104
NM_005175.2 NP_005166.1 2 - 3 172 174
GenBank mRNA AL080089.1 - 2 - 3 92 104
BG611396.1 - 82 104
CF129854.1 - 2 - 3 106 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AL080089.1, NM_001002027.1, NM_005175.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5G1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
ATP hydrolysis coupled proton transport
mitochondrial ATP synthesis coupled proton transport
transporter activity
protein binding
lipid binding
hydrogen ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0