Assay Details
| Gene Symbol: IK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006083.3 | NP_006074.2 | 2 - 3 | 195 | 108 | |
| GenBank mRNA | AF182645.1 | - | 2 - 3 | 196 | 108 | |
| AJ005579.1 | - | 2 - 3 | 85 | 108 | ||
| AK225924.1 | - | 2 - 3 | 179 | 108 | ||
| AK293626.1 | - | 2 - 3 | 153 | 108 | ||
| AK312363.1 | - | 2 - 3 | 165 | 108 | ||
| AL050296.1 | - | 2 - 3 | 161 | 108 | ||
| AY007094.1 | - | 2 - 3 | 119 | 108 | ||
| BC013005.2 | - | 2 - 3 | 150 | 3030079 | 108 | |
| BC051295.1 | - | 2 - 3 | 153 | 108 | ||
| BC071964.1 | - | 2 - 3 | 143 | 6461871 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293626.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012687.hg.1 | ENST00000434096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000417647.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000502899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000503332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000507593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000508301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000512827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000512827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000512827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000513256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000513256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000513256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000513256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000513256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000513256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000515592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000523672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000581765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000610522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | ENST00000620556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NM_006083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008863.hg.1 | NR_037428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012687.hg.1 | ENST00000434096.1 | Ensembl |

IK
Genomic Map