This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04966695_g1
See other HADHA GE Assays ›
Gene Symbol:
HADHA
Gene Name:
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases:
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
Chromosome Location:
Chr.2: 26190635 - 26244726 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04966695_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HADHA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000182.4 NP_000173.2 15 - 16 1754 93
GenBank mRNA AK293398.1 - 12 - 13 1247 93
AK299258.1 - 15 - 16 1502 93
AK313027.1 - 15 - 16 1647 93
BC009235.2 - 15 - 16 1649 2966432 93
D16480.1 - 15 - 16 1651 93
FJ626228.1 - 14 - 15 1490 93
GU727642.1 - 15 - 16 1649 93
U04627.1 - 15 - 16 1650 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HADHA

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
response to insulin
cardiolipin acyl-chain remodeling
response to drug
fatty-acyl-CoA binding
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acetyltransferase activity
enoyl-CoA hydratase activity
protein binding
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
protein complex binding
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0