This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04966775_g1
See other MOB1A GE Assays ›
Gene Symbol:
MOB1A
Gene Name:
MOB kinase activator 1A
Gene Aliases:
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B
Chromosome Location:
Chr.2: 74152598 - 74178967 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04966775_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317111.1 NP_001304040.1 1 - 2 202 110
XM_011532948.2 XP_011531250.1 1 - 2 169 110
GenBank mRNA BI825399.1 - 1 - 2 202 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

hippo signaling
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline