This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04966814_m1
See other MITD1 GE Assays ›
Gene Symbol:
MITD1
Gene Name:
microtubule interacting and trafficking domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 99161427 - 99181073 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04966814_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MITD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320417.1 NP_001307346.1 1 - 2 356 83
XM_011510583.1 XP_011508885.1 1 - 2 330 83
GenBank mRNA AK311658.1 - 1 - 2 330 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001320417.1, XM_011510583.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
cell separation after cytokinesis
negative regulation of protein binding
viral budding via host ESCRT complex
multivesicular body sorting pathway
protein binding
protein domain specific binding
phosphatidylinositol binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000289359.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000409107.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000413710.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000422537.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000438121.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000464685.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000466880.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000483721.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 ENST00000487588.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq
Affymetrix Human Clariom D Assay TC0200013603.hg.1 NM_138798.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-b5lz9:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0