This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04967399_s1
See other STARD7 GE Assays ›
Gene Symbol:
STARD7
Gene Name:
StAR related lipid transfer domain containing 7
Gene Aliases:
GTT1
Chromosome Location:
Chr.2: 96184859 - 96208846 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04967399_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020151.3 NP_064536.2 8 2403 155
XM_011511499.1 XP_011509801.1 8 2020 155
GenBank mRNA AF270647.1 - 8 2330 155
AK023234.1 - 7 1662 155
AK054615.1 - 8 1769 155
AK094925.1 - 2 1975 155
AK096666.1 - 5 1163 155
BC003028.1 - 8 2384 155
BC007894.2 - 9 2161 4134933 155
BC008894.2 - 8 2384 3833465 155
BC009998.2 - 9 2161 4128869 155
BC012774.2 - 8 2350 3842611 155
BC012793.2 - 8 2364 4080200 155
BC013279.2 - 8 2384 3830320 155
BC014076.2 - 8 2364 4127007 155
BC014274.2 - 8 2382 3677198 155
BC032106.1 - 8 2377 4643387 155
CR749695.1 - 1 718 155

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STARD7

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein targeting to mitochondrion
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0