This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04968661_s1
See other ADGRF3 GE Assays ›
Gene Symbol:
ADGRF3
Gene Name:
adhesion G protein-coupled receptor F3
Gene Aliases:
GPR113, PGR23
Chromosome Location:
Chr.2: 26308173 - 26346817 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04968661_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145168.1 NP_001138640.1 11 2708 74
NM_001145169.1 NP_001138641.1 10 2554 74
NM_001321971.1 NP_001308900.1 10 3087 74
NM_001321975.1 NP_001308904.1 11 2628 74
NM_153835.3 NP_722577.2 8 2694 74
NR_135916.1 8 2694 74
XM_011532621.2 XP_011530923.1 10 3039 74
XM_011532622.2 XP_011530924.1 10 2991 74
XM_011532623.2 XP_011530925.1 10 2983 74
XM_017003505.1 XP_016858994.1 10 3025 74
XM_017003506.1 XP_016858995.1 10 3281 74
GenBank mRNA AK122995.1 - 5 2199 74
AK293887.1 - 10 2554 74
AY140955.1 - 11 2708 74
AY255611.1 - 1 320 74
AY358172.1 - 8 2694 74
AY358214.1 - 2 1165 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRF3

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline