This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04968762_s1
See other GPR75 GE Assays ›
Gene Symbol:
GPR75
Gene Name:
G protein-coupled receptor 75
Gene Aliases:
GPRchr2, WI31133
Chromosome Location:
Chr.2: 53852913 - 53860033 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04968762_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006794.3 NP_006785.1 2 1902 102
GenBank mRNA AF072693.1 - 2 1797 102
BC067475.1 - 1 1698 6971705 102
BC067476.1 - 1 1619 6971810 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR75

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
chemokine-mediated signaling pathway
regulation of neuron death
G-protein coupled receptor activity
C-C chemokine receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline