This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04969450_g1
See other TMA7 GE Assays ›
Gene Symbol:
TMA7
Gene Name:
translation machinery associated 7 homolog
Gene Aliases:
CCDC72, HSPC016
Chromosome Location:
Chr.3: 48440276 - 48444129 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04969450_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF077202.1 - 1 - 2 54 63
AL712164.1 - 1 - 2 35 63
AY508979.1 - 1 37 63
AY794023.1 - 1 - 2 38 63
BC001102.2 - 1 26 63
BC080531.1 - 1 - 2 39 4875991 63
BU536461.1 - 1 - 2 83 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF077202.1, AY794023.1, BC080531.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0