This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970071_g1
See other ALCAM GE Assays ›
Gene Symbol:
ALCAM
Gene Name:
activated leukocyte cell adhesion molecule
Gene Aliases:
CD166, MEMD
Chromosome Location:
Chr.3: 105366713 - 105576913 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970071_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALCAM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243281.1 NP_001230210.1 13 - 14 2237 77
NM_001627.3 NP_001618.2 13 - 14 2237 77
GenBank mRNA AK054632.1 - 13 - 14 2190 77
AK095833.1 - 5 - 6 1171 77
AK127617.1 - 7 - 8 826 77
AL833702.1 - 13 - 14 2038 77
BC137096.1 - 13 - 14 1665 9021093 77
BC137097.1 - 13 - 14 1665 9021094 77
CA430528.1 - 1 - 2 77
DQ486140.1 - 13 - 14 1603 77
L38608.1 - 13 - 14 1604 77
Y10183.1 - 13 - 14 1538 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALCAM

Panther Classification:

Molecular Function -

cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
motor neuron axon guidance
retinal ganglion cell axon guidance
axon extension involved in axon guidance
neuron projection extension
receptor binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0